Product Details

SNP ID
rs139761291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10514080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCAGGAGCGCGTGGCGCAGGTCG[C/T]GGTTGGTGAGCGTGTAGATGATGGG
Phenotype
MIM: 605146
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
S1PR5 PubMed Links

Gene Details

Gene
S1PR5
Gene Name
sphingosine-1-phosphate receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166215.1 1119 Missense Mutation CAC,CGC H311R NP_001159687.1
NM_030760.4 1119 Missense Mutation CAC,CGC H311R NP_110387.1

View Full Product Details