Product Details

SNP ID
rs139888569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4047770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTTTTTAGGCGAGTCCGGCTGT[A/G]AAGTTACCGTCGGTGGCGGCCCCGG
Phenotype
MIM: 605989 MIM: 605878
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIAS4 PubMed Links

Gene Details

Gene
PIAS4
Gene Name
protein inhibitor of activated STAT 4
There are no transcripts associated with this gene.

Gene
ZBTB7A
Gene Name
zinc finger and BTB domain containing 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317990.1 2938 Silent Mutation TTC,TTT F579F NP_001304919.1
NM_015898.3 2938 Silent Mutation TTC,TTT F579F NP_056982.1
XM_005259570.4 2938 Silent Mutation TTC,TTT F629F XP_005259627.3
XM_005259571.4 2938 Silent Mutation TTC,TTT F579F XP_005259628.1

View Full Product Details