Product Details

SNP ID
rs139989420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49152328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCACCTGGCACTGGTCGCAGTG[C/T]TCACCCATGTTCTCCTCATCACAGT
Phenotype
MIM: 142705 MIM: 603144 MIM: 606936
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HRC PubMed Links

Gene Details

Gene
HRC
Gene Name
histidine rich calcium binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002152.2 2140 Silent Mutation GAA,GAG E651E NP_002143.1
XM_017026733.1 2140 UTR 3 XP_016882222.1
Gene
PPFIA3
Gene Name
PTPRF interacting protein alpha 3
There are no transcripts associated with this gene.

Gene
TRPM4
Gene Name
transient receptor potential cation channel subfamily M member 4
There are no transcripts associated with this gene.

View Full Product Details