Product Details

SNP ID
rs140022666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9528636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTCATAGGGTTTTTCTCCAGTG[C/T]GGATTCGCATGTGAATTTTAAGGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF426 PubMed Links

Gene Details

Gene
ZNF426
Gene Name
zinc finger protein 426
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300883.2 1627 Missense Mutation CAC,CGC H432R NP_001287812.1
NM_001318055.1 1627 Intron NP_001304984.1
NM_001318056.1 1627 Intron NP_001304985.1
NM_024106.2 1627 Intron NP_077011.1
XM_017027293.1 1627 Missense Mutation CAC,CGC H432R XP_016882782.1
XM_017027294.1 1627 Missense Mutation CAC,CGC H228R XP_016882783.1

View Full Product Details