Product Details

SNP ID
rs140061983
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51612277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGTACTCCGTGGTGCTTGGGGCC[A/T]CCTGGTCCTTAGGCTCCCTCGACTT
Phenotype
MIM: 604200
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
SIGLEC5 PubMed Links

Gene Details

Gene
SIGLEC5
Gene Name
sialic acid binding Ig like lectin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003830.3 1749 Missense Mutation GAG,GTG E537V NP_003821.1
XM_011527438.2 1749 UTR 3 XP_011525740.1
XM_017027419.1 1749 Missense Mutation GAG,GTG E546V XP_016882908.1

View Full Product Details