Product Details

SNP ID
rs140065601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44327780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTATAGGGTCTCCCTTCCACAT[A/G]AACCCTTTGGTGTGCTTGAAGGCGT
Phenotype
MIM: 603994
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF112 PubMed Links

Gene Details

Gene
ZNF112
Gene Name
zinc finger protein 112
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083335.1 2508 Missense Mutation CAT,TAT H799Y NP_001076804.1
NM_013380.3 2508 Missense Mutation CAT,TAT H793Y NP_037512.3

View Full Product Details