Product Details

SNP ID
rs140089736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6663771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGTGAAAGAGTGGCTGAGATTA[G/T]GCAGCCATGTGACAGATTTTGTGGG
Phenotype
MIM: 604520
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TNFSF14 PubMed Links

Gene Details

Gene
TNFSF14
Gene Name
tumor necrosis factor superfamily member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003807.4 2260 UTR 3 NP_003798.2
NM_172014.3 2260 UTR 3 NP_742011.2
XM_017027417.1 2260 UTR 3 XP_016882906.1
XM_017027418.1 2260 Intron XP_016882907.1

View Full Product Details