Product Details

SNP ID
rs140143043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1387831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGCTCCTGGCCTGCGCGGCTTC[A/C]GGATCCTTGGTCTGCGGTGAGTGCC
Phenotype
MIM: 601240 MIM: 601825
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GAMT PubMed Links

Gene Details

Gene
GAMT
Gene Name
guanidinoacetate N-methyltransferase
There are no transcripts associated with this gene.

Gene
MUM1
Gene Name
melanoma associated antigen (mutated) 1
There are no transcripts associated with this gene.

Gene
NDUFS7
Gene Name
NADH:ubiquinone oxidoreductase core subunit S7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024407.4 80 Silent Mutation AGG,CGG R13R NP_077718.3
XM_005259556.4 80 Silent Mutation AGG,CGG R13R XP_005259613.2
XM_017026768.1 80 Silent Mutation AGG,CGG R98R XP_016882257.1

View Full Product Details