Product Details

SNP ID
rs140266474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48720848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGGCGATAATTGGCTGGCAGCT[C/T]CTGCTGCTCAAGATCCCAGAGGAGG
Phenotype
MIM: 610349 MIM: 609623
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAMSTR PubMed Links

Gene Details

Gene
MAMSTR
Gene Name
MEF2 activating motif and SAP domain containing transcriptional regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130915.1 2865 Intron NP_001124387.1
NM_001297753.1 2865 Intron NP_001284682.1
NM_182574.2 2865 Intron NP_872380.1
XM_011526807.2 2865 Intron XP_011525109.1
XM_011526808.2 2865 Intron XP_011525110.1
XM_011526809.2 2865 Intron XP_011525111.1
XM_017026640.1 2865 Intron XP_016882129.1
XM_017026641.1 2865 Intron XP_016882130.1
XM_017026642.1 2865 Intron XP_016882131.1
XM_017026643.1 2865 Intron XP_016882132.1
XM_017026644.1 2865 Intron XP_016882133.1
XM_017026645.1 2865 Intron XP_016882134.1
Gene
RASIP1
Gene Name
Ras interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017805.2 2865 Missense Mutation AAG,GAG K948E NP_060275.2
XM_011527053.2 2865 Missense Mutation AAG,GAG K1059E XP_011525355.2
XM_011527054.2 2865 Intron XP_011525356.2
XM_011527055.2 2865 Missense Mutation AAG,GAG K961E XP_011525357.2
XM_017026914.1 2865 Missense Mutation AAG,GAG K1049E XP_016882403.1
XM_017026915.1 2865 Missense Mutation AAG,GAG K951E XP_016882404.1

View Full Product Details