Product Details

SNP ID
rs140294009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6368542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTCCTGCTTCCCCACCAGAGTC[C/T]GCCATGGAGAGGGACCGCTACATGA
Phenotype
MIM: 613491 MIM: 613305 MIM: 601119 MIM: 602921
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACER1 PubMed Links

Gene Details

Gene
ACER1
Gene Name
alkaline ceramidase 1
There are no transcripts associated with this gene.

Gene
ALKBH7
Gene Name
alkB homolog 7
There are no transcripts associated with this gene.

Gene
CLPP
Gene Name
caseinolytic mitochondrial matrix peptidase proteolytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006012.2 789 Silent Mutation TCC,TCT S222S NP_006003.1
Gene
PSPN
Gene Name
persephin
There are no transcripts associated with this gene.

View Full Product Details