Product Details

SNP ID
rs140295044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1556577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAGTGACCGCGAACACCGGCTC[C/T]TTGTCGCGCCCGGGCGTCACGATGT
Phenotype
MIM: 611009
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEX3D PubMed Links

Gene Details

Gene
MEX3D
Gene Name
mex-3 RNA binding family member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174118.1 942 Silent Mutation AAA,AAG K314K NP_001167589.1
NM_203304.3 942 Silent Mutation AAA,AAG K314K NP_976049.3
XM_017026811.1 942 Silent Mutation AAA,AAG K80K XP_016882300.1

View Full Product Details