Product Details

SNP ID
rs140295213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57572643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGCTGAATGAGGCCACACTTTA[A/G]GCTAAATGATTTCCCACACTGGCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF416 PubMed Links

Gene Details

Gene
ZNF416
Gene Name
zinc finger protein 416
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017879.1 1432 Silent Mutation CTA,TTA L421L NP_060349.1
XM_005259051.3 1432 Silent Mutation CTA,TTA L349L XP_005259108.1

View Full Product Details