Product Details

SNP ID
rs140304059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39329052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTTTTTACTCCCTGCATACATC[A/G]TCTGTTGTTCCGGCTTGCAGCCTGC
Phenotype
MIM: 604104 MIM: 612807
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GMFG PubMed Links

Gene Details

Gene
GMFG
Gene Name
glia maturation factor gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301008.1 415 Missense Mutation ACG,ATG T61M NP_001287937.1
NM_004877.3 415 Missense Mutation ACG,ATG T102M NP_004868.1
Gene
LRFN1
Gene Name
leucine rich repeat and fibronectin type III domain containing 1
There are no transcripts associated with this gene.

View Full Product Details