Product Details

SNP ID
rs140377964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33081223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCGCGGGACAGTGACAGCGAAGA[A/G]GATCTGGTCAGCTATGGGACCGGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPATCH1 PubMed Links

Gene Details

Gene
GPATCH1
Gene Name
G-patch domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018025.2 59 Silent Mutation GAA,GAG E10E NP_060495.2
XM_006723255.3 59 Silent Mutation GAA,GAG E10E XP_006723318.1

View Full Product Details