Product Details

SNP ID
rs140399351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42208358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTCTCCTCACCCTCTCCAACAC[A/G]ACCCCCAGGCCAGACCTCTGCCTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEDD2 PubMed Links

Gene Details

Gene
DEDD2
Gene Name
death effector domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270614.1 Intron NP_001257543.1
NM_001270615.1 Intron NP_001257544.1
NM_133328.3 Intron NP_579874.1
XM_011526569.1 Intron XP_011524871.1
XM_011526571.1 Intron XP_011524873.1
XM_011526572.1 Intron XP_011524874.1
XM_017026402.1 Intron XP_016881891.1
XM_017026403.1 Intron XP_016881892.1

View Full Product Details