Product Details

SNP ID
rs140412357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54118577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTTCTCCTTTCCTACAGTTTG[C/G]TGAGATTATGATGAAGATTGAGGAG
Phenotype
MIM: 606419 MIM: 609519
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRPF31 PubMed Links

Gene Details

Gene
PRPF31
Gene Name
pre-mRNA processing factor 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015629.3 578 Missense Mutation GCT,GGT A61G NP_056444.3
XM_006723137.3 578 Missense Mutation GCT,GGT A61G XP_006723200.1
Gene
TFPT
Gene Name
TCF3 fusion partner
There are no transcripts associated with this gene.

View Full Product Details