Product Details
- SNP ID
-
rs140414099
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:15052207 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACCTATCTTCTCTTGACTCCAAGG[A/G]TCAGACAAGGGTGCTGAGAGCCGGG
- Phenotype
-
MIM: 605848
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CASP14
PubMed Links
Gene Details
- Gene
- CASP14
- Gene Name
- caspase 14
View Full Product Details