Product Details

SNP ID
rs140472122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49907174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAACTCACAATCTAACAGCGAGA[C/T]GAGGCACAAACGGCTAGCACAGGAT
Phenotype
MIM: 609742 MIM: 605815
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL4I1 PubMed Links

Gene Details

Gene
IL4I1
Gene Name
interleukin 4 induced 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258017.1 2837 Intron NP_001244946.1
NM_001258018.1 2837 Intron NP_001244947.1
NM_152899.1 2837 Intron NP_690863.1
NM_172374.2 2837 Intron NP_758962.1
Gene
NUP62
Gene Name
nucleoporin 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193357.1 2837 UTR 3 NP_001180286.1
NM_012346.4 2837 UTR 3 NP_036478.2
NM_016553.4 2837 UTR 3 NP_057637.2
NM_153718.3 2837 UTR 3 NP_714940.1
NM_153719.3 2837 UTR 3 NP_714941.1

View Full Product Details