Product Details

SNP ID
rs140545844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35049332 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGCTCCAGACCCAAGGTGGCAG[C/T]TCTCACTGCGGGGACCCTGCTACTT
Phenotype
MIM: 142440 MIM: 600235
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HPN PubMed Links

Gene Details

Gene
HPN
Gene Name
hepsin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002151.2 258 Missense Mutation GCT,GTT A20V NP_002142.1
NM_182983.2 258 Missense Mutation GCT,GTT A20V NP_892028.1
XM_005258838.4 258 Missense Mutation GCT,GTT A20V XP_005258895.2
XM_006723181.3 258 Missense Mutation GCT,GTT A20V XP_006723244.2
XM_017026731.1 258 Missense Mutation GCT,GTT A20V XP_016882220.1
XM_017026732.1 258 UTR 5 XP_016882221.1
Gene
HPN-AS1
Gene Name
HPN antisense RNA 1
There are no transcripts associated with this gene.

Gene
SCN1B
Gene Name
sodium voltage-gated channel beta subunit 1
There are no transcripts associated with this gene.

View Full Product Details