Product Details
- SNP ID
-
rs140789422
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:54162764 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACATTTCCTGGCCCAGGACAGTCT[C/T]CCAGCACTGAAGAAGGAAGAAATAT
- Phenotype
-
MIM: 604910
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CNOT3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9636146] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CNOT3
- Gene Name
- CCR4-NOT transcription complex subunit 3
There are no transcripts associated with this gene.
- Gene
- LENG1
- Gene Name
- leukocyte receptor cluster member 1
There are no transcripts associated with this gene.
- Gene
- TMC4
- Gene Name
- transmembrane channel like 4
View Full Product Details