Product Details

SNP ID
rs140821686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49336988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCACCATGGGCATCGCCCTCCT[A/G]GGTTGTGTGGGGGCCCTCAAGGAGC
Phenotype
MIM: 151523 MIM: 607972 MIM: 601729
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD37 PubMed Links

Gene Details

Gene
CD37
Gene Name
CD37 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040031.1 341 Silent Mutation CTA,CTG L6L NP_001035120.1
NM_001774.2 341 Silent Mutation CTA,CTG L74L NP_001765.1
XM_005259435.3 341 Silent Mutation CTA,CTG L74L XP_005259492.1
XM_005259436.3 341 Silent Mutation CTA,CTG L6L XP_005259493.1
XM_011527542.2 341 Silent Mutation CTA,CTG L6L XP_011525844.1
XM_011527543.2 341 Silent Mutation CTA,CTG L6L XP_011525845.1
XM_011527544.2 341 Silent Mutation CTA,CTG L6L XP_011525846.1
XM_017027513.1 341 Silent Mutation CTA,CTG L6L XP_016883002.1
Gene
SLC6A16
Gene Name
solute carrier family 6 member 16
There are no transcripts associated with this gene.

Gene
TEAD2
Gene Name
TEA domain transcription factor 2
There are no transcripts associated with this gene.

View Full Product Details