Product Details

SNP ID
rs140824256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57124071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGGCAGTCAGAGCTCTGCCTGCA[C/G]TGTGACCATGGACAGCTCTCCAAAG
Phenotype
MIM: 609546
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
USP29 PubMed Links

Gene Details

Gene
USP29
Gene Name
ubiquitin specific peptidase 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020903.2 272 UTR 5 NP_065954.1

View Full Product Details