Product Details

SNP ID
rs140839998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12956978 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCCAGCTCCCGCAGCTGCTCCG[C/G]CGACGGCCATAACGAACCGGGGACC
Phenotype
MIM: 605162 MIM: 600061
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GADD45GIP1 PubMed Links

Gene Details

Gene
GADD45GIP1
Gene Name
GADD45G interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052850.3 277 Missense Mutation CCG,GCG P79A NP_443082.2
Gene
RAD23A
Gene Name
RAD23 homolog A, nucleotide excision repair protein
There are no transcripts associated with this gene.

View Full Product Details