Product Details

SNP ID
rs140844969
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47029864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACACAGAAGTGGATTTGCTAACTC[A/G]TATGATAATTCCATGTTAACTTATT
Phenotype
MIM: 603346
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NPAS1 PubMed Links

Gene Details

Gene
NPAS1
Gene Name
neuronal PAS domain protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321086.1 Intron NP_001308015.1
NM_002517.3 Intron NP_002508.2
XM_006723231.2 Intron XP_006723294.1
XM_011526997.1 Intron XP_011525299.1
XM_011526999.1 Intron XP_011525301.1
XM_011527001.1 Intron XP_011525303.1
XM_011527002.1 Intron XP_011525304.1
XM_011527006.1 Intron XP_011525308.1
XM_011527007.1 Intron XP_011525309.1
XM_017026841.1 Intron XP_016882330.1
XM_017026842.1 Intron XP_016882331.1
XM_017026843.1 Intron XP_016882332.1
XM_017026844.1 Intron XP_016882333.1

View Full Product Details