Product Details

SNP ID
rs140857061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45884527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCAATGGGCGAGGGCACACCAGG[A/G]GTCTCAGCGGAGTACGGGCCGCCGG
Phenotype
MIM: 615331
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRF2BP1 PubMed Links

Gene Details

Gene
IRF2BP1
Gene Name
interferon regulatory factor 2 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015649.2 1644 Silent Mutation ACC,ACT T416T NP_056464.1
Gene
MYPOP
Gene Name
Myb related transcription factor, partner of profilin
There are no transcripts associated with this gene.

View Full Product Details