Product Details

SNP ID
rs140889768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52766425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGAAAACCTTTTCACATTCTTCA[A/T]ATTTGTAAGGTTGCTCTCCAGTATG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF600 PubMed Links

Gene Details

Gene
ZNF600
Gene Name
zinc finger protein 600
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321866.1 Intron NP_001308795.1
NM_001321867.1 Intron NP_001308796.1
NM_198457.3 Intron NP_940859.3
XM_017026390.1 Intron XP_016881879.1
XM_017026391.1 Intron XP_016881880.1
XM_017026392.1 Intron XP_016881881.1

View Full Product Details