Product Details

SNP ID
rs140965080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21809340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATTAAAGACTTTGCCACATTCTT[C/T]ACATGTGTAGGGTTTCTCTCCAGTA
Phenotype
MIM: 603972
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF43 PubMed Links

Gene Details

Gene
ZNF43
Gene Name
zinc finger protein 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256648.1 6525 Intron NP_001243577.1
NM_001256649.1 6525 Missense Mutation AAA,GAA K227E NP_001243578.1
NM_001256650.1 6525 Intron NP_001243579.1
NM_001256651.1 6525 Intron NP_001243580.1
NM_001256653.1 6525 Intron NP_001243582.1
NM_001256654.1 6525 Missense Mutation AAA,GAA K168E NP_001243583.1
NM_003423.3 6525 Missense Mutation AAA,GAA K233E NP_003414.2
XM_011528257.2 6525 Intron XP_011526559.1
XM_011528259.2 6525 Missense Mutation AAA,GAA K227E XP_011526561.1
XM_017027207.1 6525 Intron XP_016882696.1
XM_017027208.1 6525 Intron XP_016882697.1
XM_017027209.1 6525 Intron XP_016882698.1
XM_017027210.1 6525 Intron XP_016882699.1
XM_017027211.1 6525 Missense Mutation AAA,GAA K227E XP_016882700.1
XM_017027212.1 6525 Intron XP_016882701.1
XM_017027213.1 6525 Intron XP_016882702.1
XM_017027214.1 6525 Intron XP_016882703.1
XM_017027215.1 6525 Intron XP_016882704.1
XM_017027216.1 6525 Intron XP_016882705.1

View Full Product Details