Product Details

SNP ID
rs140987549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3977285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGCCGGCCACCTGGGACTCCTC[A/G]AACACGTGGCCCCGCTTCCTGTTCA
Phenotype
MIM: 603289 MIM: 130610
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DAPK3 PubMed Links

Gene Details

Gene
DAPK3
Gene Name
death associated protein kinase 3
There are no transcripts associated with this gene.

Gene
EEF2
Gene Name
eukaryotic translation elongation factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001961.3 2396 Silent Mutation TTC,TTT F771F NP_001952.1
Gene
SNORD37
Gene Name
small nucleolar RNA, C/D box 37
There are no transcripts associated with this gene.

View Full Product Details