Product Details

SNP ID
rs141028740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48333314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAACCCGGGGGTGGGTACAATC[C/T]CATGCTGGGGGTCAGGGCCTGCAGA
Phenotype
MIM: 602335
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EMP3 PubMed Links

Gene Details

Gene
EMP3
Gene Name
epithelial membrane protein 3
There are no transcripts associated with this gene.

Gene
TMEM143
Gene Name
transmembrane protein 143
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303538.1 1497 Missense Mutation AGA,GGA R394G NP_001290467.1
NM_001303539.1 1497 Missense Mutation AGA,GGA R364G NP_001290468.1
NM_001303540.1 1497 Missense Mutation AGA,GGA R329G NP_001290469.1
NM_018273.3 1497 Missense Mutation AGA,GGA R429G NP_060743.2

View Full Product Details