Product Details
- SNP ID
-
rs141106192
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:14028359 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTTCACCTGCGGGGGCTCCCGGT[G/T]GAGACGATCAGACATCCTGGCCCAC
- Phenotype
-
MIM: 605350
MIM: 606855
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IL27RA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79665112] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IL27RA
- Gene Name
- interleukin 27 receptor subunit alpha
There are no transcripts associated with this gene.
- Gene
- RLN3
- Gene Name
- relaxin 3
View Full Product Details