Product Details

SNP ID
rs141146690
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5867538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGTCGCCATGCTGTCCTGGCAG[C/G]GGGACCCATTGGGAGCCCCGGTGAC
Phenotype
MIM: 136835
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
FUT5 PubMed Links

Gene Details

Gene
FUT5
Gene Name
fucosyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002034.2 276 Missense Mutation CCC,CGC P63R NP_002025.2
Gene
LOC101928844
Gene Name
uncharacterized LOC101928844
There are no transcripts associated with this gene.

View Full Product Details