Product Details
- SNP ID
-
rs141166390
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:54594876 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCCATCACCTGGGAACACACAGG[A/G]CGGTATCGCTGTTTCTACGGTAGCC
- Phenotype
-
MIM: 604810
MIM: 604812
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LILRA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16989288] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LILRA1
- Gene Name
- leukocyte immunoglobulin like receptor A1
- Gene
- LILRA2
- Gene Name
- leukocyte immunoglobulin like receptor A2
There are no transcripts associated with this gene.
View Full Product Details