Product Details
- SNP ID
-
rs141183673
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:12701385 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCCGCCAGCCTCTCCTTGAGCAG[C/T]GGCGGGAATTGCTCAGAGAACTGCT
- Phenotype
-
MIM: 609074
MIM: 603002
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FBXW9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16978628] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FBXW9
- Gene Name
- F-box and WD repeat domain containing 9
There are no transcripts associated with this gene.
- Gene
- SNORD135
- Gene Name
- small nucleolar RNA, C/D box 135
There are no transcripts associated with this gene.
- Gene
- SNORD41
- Gene Name
- small nucleolar RNA, C/D box 41
There are no transcripts associated with this gene.
- Gene
- TNPO2
- Gene Name
- transportin 2
View Full Product Details