Product Details

SNP ID
rs141266080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10252596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAAAGTCGAGCTCCCGGTACCCA[C/T]TCATCGACGCCCAGTGCAGGCCTGG
Phenotype
MIM: 611823
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL4 PubMed Links

Gene Details

Gene
MRPL4
Gene Name
mitochondrial ribosomal protein L4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015956.2 324 Missense Mutation ACT,ATT T57I NP_057040.2
NM_146387.1 324 Missense Mutation ACT,ATT T57I NP_666499.1
NM_146388.1 324 Missense Mutation ACT,ATT T57I NP_666500.1
XM_011528045.2 324 Missense Mutation ACT,ATT T57I XP_011526347.1

View Full Product Details