Product Details

SNP ID
rs141272910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55359026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCTTGGACTCCTTCTTGGTCAC[A/G]GTGAAGGGCGACTCCGGGGACTTGG
Phenotype
MIM: 147681
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COX6B2 PubMed Links

Gene Details

Gene
COX6B2
Gene Name
cytochrome c oxidase subunit 6B2
There are no transcripts associated with this gene.

Gene
FAM71E2
Gene Name
family with sequence similarity 71 member E2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145402.1 2028 Silent Mutation ACC,ACT T614T NP_001138874.1
Gene
IL11
Gene Name
interleukin 11
There are no transcripts associated with this gene.

View Full Product Details