Product Details

SNP ID
rs141330883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41837217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGCCTGGAGGATGCTGCACCAA[C/G]AATCTGAGACTTCTCTAAGATGTTG
Phenotype
MIM: 614806
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DMRTC2 PubMed Links

Gene Details

Gene
DMRTC2
Gene Name
DMRT like family C2
There are no transcripts associated with this gene.

Gene
LYPD4
Gene Name
LY6/PLAUR domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291419.1 911 Missense Mutation CTT,GTT L188V NP_001278348.1
NM_173506.6 911 Missense Mutation CTT,GTT L223V NP_775777.3
XM_005258543.3 911 Missense Mutation CTT,GTT L236V XP_005258600.1
XM_005278383.4 911 Missense Mutation CTT,GTT L236V XP_005278440.1
XM_005278385.4 911 Missense Mutation CTT,GTT L236V XP_005278442.1
XM_005278387.4 911 Intron XP_005278444.1
XM_005278388.3 911 Missense Mutation CTT,GTT L174V XP_005278445.1
XM_006723035.2 911 Missense Mutation CTT,GTT L232V XP_006723098.1
XM_011526483.2 911 Missense Mutation CTT,GTT L236V XP_011524785.1
XM_011526484.2 911 Missense Mutation CTT,GTT L236V XP_011524786.1

View Full Product Details