Product Details

SNP ID
rs141361008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45488937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCGGGAGGTGATCTGGTGGGACA[A/G]ACGTTCCGTCTGCTCCCGAGTCAGG
Phenotype
MIM: 603183
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPM1N PubMed Links

Gene Details

Gene
PPM1N
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1N (putative)
There are no transcripts associated with this gene.

Gene
RTN2
Gene Name
reticulon 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005619.4 1521 Silent Mutation CTG,TTG L431L NP_005610.1
NM_206900.2 1521 Silent Mutation CTG,TTG L358L NP_996783.1
NM_206901.2 1521 Silent Mutation CTG,TTG L91L NP_996784.1

View Full Product Details