Product Details

SNP ID
rs141412861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41967697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGCGCAGGGCCACGTCCATGCC[A/G]GGGCAGTAGGACAGGAAGGCAGCCA
Phenotype
MIM: 182350 MIM: 604925
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP1A3 PubMed Links

Gene Details

Gene
ATP1A3
Gene Name
ATPase Na+/K+ transporting subunit alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256213.1 3125 Silent Mutation CCC,CCT P973P NP_001243142.1
NM_001256214.1 3125 Silent Mutation CCC,CCT P975P NP_001243143.1
NM_152296.4 3125 Silent Mutation CCC,CCT P962P NP_689509.1
Gene
RABAC1
Gene Name
Rab acceptor 1
There are no transcripts associated with this gene.

View Full Product Details