Product Details

SNP ID
rs141471799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1397400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGCGTGATCATCTGTGGGAAGG[C/T]GTAGTAGCGGCAGTCGGCCGGTGGG
Phenotype
MIM: 601240 MIM: 601825
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAMT PubMed Links

Gene Details

Gene
GAMT
Gene Name
guanidinoacetate N-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000156.5 764 Missense Mutation ACC,GCC T224A NP_000147.1
NM_138924.2 764 Intron NP_620279.1
Gene
NDUFS7
Gene Name
NADH:ubiquinone oxidoreductase core subunit S7
There are no transcripts associated with this gene.

View Full Product Details