Product Details
- SNP ID
-
rs141578342
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:41354712 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGCAGAGTCCCTCAGCACTCCAC[A/G]CCGTAGCGGTCGAAGTTGCGGAGCA
- Phenotype
-
MIM: 611951
MIM: 190180
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
B9D2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1800468] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- B9D2
- Gene Name
- B9 protein domain 2
- Gene
- TGFB1
- Gene Name
- transforming growth factor beta 1
- Gene
- TMEM91
- Gene Name
- transmembrane protein 91
There are no transcripts associated with this gene.
View Full Product Details