Product Details

SNP ID
rs141633091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55226735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGGCCTGGTCCATGGGGAGCGA[C/T]AAAGAACCTTCTGCTGTCATTAACC
Phenotype
MIM: 610875
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6804 PubMed Links

Gene Details

Gene
MIR6804
Gene Name
microRNA 6804
There are no transcripts associated with this gene.

Gene
PPP6R1
Gene Name
protein phosphatase 6 regulatory subunit 1
There are no transcripts associated with this gene.

Gene
TMEM86B
Gene Name
transmembrane protein 86B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173804.4 1423 UTR 3 NP_776165.3
XM_017026554.1 1423 UTR 3 XP_016882043.1

View Full Product Details