Product Details
- SNP ID
-
rs141828389
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57157243 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACACCCTACACAGCAGTCTGACAA[C/T]TTTATTCCATTTCAAGATGTGTCAG
- Phenotype
-
MIM: 611168
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DUXA
PubMed Links
Gene Details
- Gene
- DUXA
- Gene Name
- double homeobox A
View Full Product Details