Product Details
- SNP ID
-
rs141829596
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:52115079 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATGTGAACTATGCCTGAATACCTT[A/G]CCACATTCATCACATTTATATGGTT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF616
PubMed Links
Gene Details
- Gene
- ZNF616
- Gene Name
- zinc finger protein 616
View Full Product Details