Product Details
- SNP ID
-
rs141866022
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:37564726 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCAGAGTAAGTTCTGAGCCACGAC[C/T]AAAGGCCCTCCCACATTCCTTACAT
- Phenotype
-
MIM: 613903
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF540
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16973890] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF540
- Gene Name
- zinc finger protein 540
- Gene
- ZNF571
- Gene Name
- zinc finger protein 571
- Gene
- ZNF571-AS1
- Gene Name
- ZNF571 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details