Product Details

SNP ID
rs141888879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17227193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAGGCCCCACCCAGTGCCCGACT[A/G]TGAGCTGTGAGTGTCCCCCATGTGA
Phenotype
MIM: 132880 MIM: 610675
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NR2F6 PubMed Links

Gene Details

Gene
NR2F6
Gene Name
nuclear receptor subfamily 2 group F member 6
There are no transcripts associated with this gene.

Gene
OCEL1
Gene Name
occludin/ELL domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024578.2 490 Missense Mutation TAT,TGT Y149C NP_078854.1
XM_005260079.2 490 Missense Mutation TAT,TGT Y93C XP_005260136.1
XM_006722899.3 490 Missense Mutation TAT,TGT Y149C XP_006722962.1
XM_017027306.1 490 Missense Mutation TAT,TGT Y93C XP_016882795.1
Gene
USE1
Gene Name
unconventional SNARE in the ER 1
There are no transcripts associated with this gene.

View Full Product Details