Product Details

SNP ID
rs141962525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41194676 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGAGTTCAGCGGCCGGGGAACC[A/G]TAGCGATGCTGGAAGGGACTTTTGA
Phenotype
MIM: 611529
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP2S1 PubMed Links

Gene Details

Gene
CYP2S1
Gene Name
cytochrome P450 family 2 subfamily S member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030622.7 368 Missense Mutation ATA,GTA I104V NP_085125.1

View Full Product Details