Product Details

SNP ID
rs141990164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56223717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTGGACGCATCTGGTTCACCG[A/C]GGAGGCCCGTTGGCTGGACACGTCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZSCAN5A PubMed Links

Gene Details

Gene
ZSCAN5A
Gene Name
zinc finger and SCAN domain containing 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322061.1 851 Missense Mutation GCG,TCG A221S NP_001308990.1
NM_001322062.1 851 Intron NP_001308991.1
NM_001322064.1 851 Missense Mutation GCG,TCG A168S NP_001308993.1
NM_001322065.1 851 Missense Mutation GCG,TCG A168S NP_001308994.1
NM_001322066.1 851 Missense Mutation GCG,TCG A168S NP_001308995.1
NM_001322067.1 851 Missense Mutation GCG,TCG A168S NP_001308996.1
NM_001322068.1 851 Missense Mutation GCG,TCG A168S NP_001308997.1
NM_001322069.1 851 Missense Mutation GCG,TCG A168S NP_001308998.1
NM_001322070.1 851 Missense Mutation GCG,TCG A168S NP_001308999.1
NM_001322072.1 851 Missense Mutation GCG,TCG A168S NP_001309001.1
NM_001322073.1 851 Missense Mutation GCG,TCG A168S NP_001309002.1
NM_001322074.1 851 Missense Mutation GCG,TCG A168S NP_001309003.1
NM_001322075.1 851 Missense Mutation GCG,TCG A168S NP_001309004.1
NM_001322076.1 851 Missense Mutation GCG,TCG A168S NP_001309005.1
NM_001322077.1 851 Missense Mutation GCG,TCG A51S NP_001309006.1
NM_001322078.1 851 Missense Mutation GCG,TCG A38S NP_001309007.1
NM_024303.2 851 Intron NP_077279.1
XM_005259254.1 851 Intron XP_005259311.1
XM_006723377.3 851 Missense Mutation GCG,TCG A244S XP_006723440.1
XM_011527308.1 851 Intron XP_011525610.1
XM_017027298.1 851 Missense Mutation GCG,TCG A244S XP_016882787.1
XM_017027299.1 851 Missense Mutation GCG,TCG A168S XP_016882788.1

View Full Product Details