Product Details

SNP ID
rs142006433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49528619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGACGCTCCCCATGATGACGCCCA[C/T]GGGAACTTCCAGTACGACCATGAGG
Phenotype
MIM: 601437
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCGRT PubMed Links

Gene Details

Gene
FCGRT
Gene Name
Fc fragment of IgG receptor and transporter
There are no transcripts associated with this gene.

Gene
RCN3
Gene Name
reticulocalbin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020650.2 868 Silent Mutation CAC,CAT H49H NP_065701.2
XM_005259089.3 868 UTR 5 XP_005259146.1
XM_011527143.1 868 Silent Mutation CAC,CAT H49H XP_011525445.1
XM_017027023.1 868 Silent Mutation CAC,CAT H49H XP_016882512.1

View Full Product Details