Product Details

SNP ID
rs142015120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51003204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTCTGGAGCAGGGCTCAGCACC[A/C]CCAGACACCACGCCTGCCAAGGTTC
Phenotype
MIM: 602673 MIM: 605644 MIM: 605504
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KLK10 PubMed Links

Gene Details

Gene
KLK10
Gene Name
kallikrein related peptidase 10
There are no transcripts associated with this gene.

Gene
KLK8
Gene Name
kallikrein related peptidase 8
There are no transcripts associated with this gene.

Gene
KLK9
Gene Name
kallikrein related peptidase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012315.1 747 Silent Mutation GGG,GGT G220G NP_036447.1

View Full Product Details