Product Details
- SNP ID
-
rs142024526
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:52274435 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCATTCCTTGTATTTGTGGTGATGC[C/T]GGCGGAAACAAACCTAGTGCGTTAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR643
PubMed Links
Gene Details
- Gene
- MIR643
- Gene Name
- microRNA 643
There are no transcripts associated with this gene.
- Gene
- ZNF766
- Gene Name
- zinc finger protein 766
View Full Product Details